Would you like to log in again?
Red Cabbage pH Indicator Kit
Supplier: UNIVERSE OF SCIENCE INC SE
2150-B
470145-176CS 305.2 USD InStock
470145-176
Red Cabbage pH Indicator Kit
Educational Materials Chemistry Educational Materials Acids and Bases Learning Activities
Use Red Cabbage Extract as a Natural Acid-Base Indicator
No need to be intimidated by acid-base reactions with this classroom safe pH activity. Use red cabbage extract, a natural acid-base indicator, to illustrate common reactions. Show students how to use pH charts and distinguish between chemical components. Made of all food grade materials, clean up and disposal of this project is quick and easy. Materials in this kit include 20 g red cabbage extract, plastic beaker, pipet, pH chart, and instructions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty