Would you like to log in again?
Octagon Spinbar Magnetic Stirring Bars
Supplier: BEL-ART PRODUCTS SE
371106006 F37110-0238_SE 37110-5018_SE 371106004
470145-272CS 262.35 USD InStock
470145-272 470121-044 470145-280 470145-278
Octagon Spinbar Magnetic Stirring Bars
Stirrers Magnetic Stirring Bars
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape.
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low friction coating; durable and inert
  • Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars
  • Autoclavable
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
Pivot ring aids in reducing friction and chattering (470145-282 does not have a pivot ring).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty