Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Flying Model Rocket Kit - Wizard™

Supplier: ESTES INDUSTRIES INC SE
1292
470145-390CS 412 USD InStock
470145-390
Flying Model Rocket Kit - Wizard™
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Quick to assemble with high performance. 12"L w/ 30" streamer.

  • Fins: Die-cut balsa
  • Weight without engine: 0.79 oz.,
  • Engine pptions: A8-3, B4-4, B6-4, B6-6, C6-5, C6-7
  • Likely altitude: 200 - 1600'
  • Min. dia. launch site: 50 - 400'

Needs Painting: Yes

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty