Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Ni-MH Rechargeable Batteries and Chargers
Supplier: RAYOVAC CORPORATION
PL715-4GENE PS204GENE PS335-4BGENE PL714-2GENE PS134-4BGENE PL713-2GENE PL1604-1GENE PL724-4GENE
470145-492CS 144.95 USD InStock
470145-492 470145-494 470317-614 470145-490 470039-372 470102-432 470102-430 470317-640
Ni-MH Rechargeable Batteries and Chargers
Batteries
Spend less on batteries by charging them over and over, and help the envrionment at the same time.
  • Supplied already charged
  • Work in all battery chargers
  • Long lasting and dependable
  • Great for digital devices
Nickel Metal Hydride or NiMH batteries are considered better than NiCad batteries as they are less likely to exhibit the “memory” effect, have a higher storage capacity (+25%), and there is no toxic cadmium to dispose of when they finally wear out. NiMH batteries last an average 6,000 hours and can keep 80% of their charge at -20°. Batteries are rechargeable using the chargers sold separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty