Would you like to log in again?
Fizz Wizard and Jammin’ Jelly Reaction
Supplier: UNIVERSE OF SCIENCE INC SE
2100-B
470145-532CS 388.3 USD InStock
470145-532
Fizz Wizard and Jammin’ Jelly Reaction
Educational Materials Chemistry Educational Materials Consumer Chemistry Learning Activities
Chemistry experiments you can drink and eat. Make soda pop and make it change colors. Pull tasty jelly things from a solution. Make Polymer Pudding. Learn about chemical reactions, acidbase reactions, indicators, pH and gels. Note: This kit contains food grade materials. Do not attempt these experiments with any other chemical samples. Kit contains: Magic Color Crystals (cabbage extract), sodium bicarbonate, citric acid, calcium acetate, sodium alginate, pipet, spoon, small scoops, cups, stirrers and instructions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty