Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Boreal Polarizing Microscopes
Supplier: VWR
Boreal
POL-290 5790200
470145-556EA 1012.45 USD InStock
470145-556 470012-234
Boreal Polarizing Microscopes
Microscopes
Ideal for introductory thin section microscopy.
  • Height: 15 1/2"; Footprint: 7" x 8"; Weight: 4 lbs., 13 oz.
  • Widefield 10X/18 eyepiece with cross reticle
  • 5 1/4" stage diameter
  • Halogen illumination with intensity control
  • Boreal Lifetime Warranty
The Boreal Polarizing Microscope: a full slate of features and a low cost make our polarizing microscope the perfect choice. It has a 45° inclined monocular head that is rotatable 360°. A slide in analyzer with both a 4/λ plate or a λ plate and a Bertrand lens come with the scope. This microscope also has achromatic objectives EA4X, EA10X, and EA40X; a working table that rotates from 0–360° with graduations of 1°; and coaxial coarse and fine focusing.
Boreal Advanced Polarizing Microscope: the improved design of this advanced microscope allows you to swing each polarizer completely out of the light path. You can also adjust the optical orientation of the substage polarizer by use of a 360° rotatable ring. High quality achromatic objectives (EA4X, EA10X, and EA40X), a Bertrand lens, and bright, adjustable halogen illumination provide crystal clear views.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty