Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Boreal Science Student Stereomicroscopes
Supplier: VWR
Boreal
5540400 5572803 2/CS 5744900 S-20-P
470012-230EA 350.65 USD InStock
470012-230 470145-720 470145-578 470145-710 470145-564
Boreal Science Student Stereomicroscopes
Microscopes
These stereomicroscopes have all the quality features and versatility of larger stereo microscopes in a compact size.
  • Adjustable Interpupillary Distance
  • Interchangeable Stage Plates: Translucent and Black/White
  • Lifetime Manufacturer's Warranty
  • Diopter Adjustment on One Eyetube
Features include 45° inclined viewing heads, post mounts that adjust the head up or down to accommodate larger objects on stage, and an easy-adjust collar for precise control of focus tension.
Rugged, die-cast metal construction and a reagent resistant finish make these stereomicroscopes durable.
Ordering information: This microscope includes removable soft eyecups and vinyl dust cover.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty