Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Boreal Science Digital Compound Microscopes
Supplier: VWR
Boreal
1100400200075 5584700 DC5-169-ASC
470145-726EA 730.05 USD InStock
470145-726 470145-722 470222-452
Boreal Science Digital Compound Microscopes
Microscopes
In these digital compound instruments, a video camera is built into the traditional microscope chassis.
  • Motic image editing software included
  • Smartboard compatible
  • One-year manufacturer's warranty on camera
  • Boreal lifetime warranty on microscope chassis
  • VAC: 100 - 120
These rugged digital microscopes offer high resolutions. One power supply operates both the microscope and camera. The included, user-friendly Motic image editing software lets you capture images in JPG, TIFF, MIG, and BMP formats and movies in AVI format.
Digital and Digital2000 microscopes connect to a computer via a USB port. Each microscope comes with all-connecting cables.
Ordering information: This product includes a manufacturer's warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty