Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Boreal Advanced Compound Microscope
Supplier: VWR
Boreal
5584400 5791102 5584500
470145-590EA 580.85 USD InStock
470145-590 470177-238 470145-732
Boreal Advanced Compound Microscope
Microscopes
Exceptional Quality at an Outstanding Value
  • Rack-and-Pinion Focus Guide
  • 360° Rotatable Head for Viewing from any Angle
  • Long-Lasting LED Illumination
  • Height: 141/2"
  • Lifetime Manufacturer's Warranty
Multiple features make this a practical choice for the classroom including 4x, 10x, 40x and 100x objectives, built-in mechanical stage and corded or cordless LED illumination. Designed to withstand years of wear and tear, it features all metal construction. A reinforced joint between head and eyetube prevent stripping and recessed set screws prevent tampering. Includes Boreal Lifetime Warranty.
Two people can view a specimen simultaneously, or one viewer can add a micro-video camera with the two 10x wide-field eyepieces on this version of the high-quality scope.
Ordering information: This product includes a manufacturer's warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty