Would you like to log in again?
Weather Gauge
Supplier: Skywill Far East Industries
6360300
470145-752BX 2079.4 USD InStock
470145-752
Weather Gauge
Educational Materials Earth Science Educational Materials Geology Learning Activities
Multifunction Capability in One Affordable Instrument
  • Can be mounted on a vertical support
The plastic gauge simultaneously indicates wind speed from 0–60 mph; wind direction; rainfall up to 4", up to 20" cumulatively; and temperature, -40°F to +100°F. The gauge spins to indicate wind direction when mounted on a vertical support. Size: 7"L x 6 3/4"W x 1"H.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty