Would you like to log in again?
Determination of Dissolved Oxygen Concentration
Supplier: ALDON CORP SE
IS9750
470146-436EA 40.75 USD InStock
470146-436
Determination of Dissolved Oxygen Concentration
Educational Materials Chemistry Educational Materials Mixtures and Solutions Learning Activities
With this kit, students determine dissolved oxygen concentration without a meter.
  • Great for field studies
  • Perfect for environmental or chemistry courses
  • All required chemicals included
  • Kit features a teacher's guide
  • Students determine dissolved oxygen concentration without a meter
  • Aligned with GLOBE protocols
Students perform a simple titration to determine the dissolved oxygen concentration in their fresh water sample. The titration uses an easily identifiable color transition to indicate the end of the analysis.
The kit is perfect for environmental or chemistry courses and contains everything an analyst would need to perform a water analysis either in the laboratory setting or out in the field. It also contains enough material to perform 40 tests.
Ordering information: This kit includes pipettes, a reaction vessel, a collection bottle, a measuring tube, a titrator, manganese chloride reagent, alkaline iodide reagent, sulfuric acid solution, sodium thiosulfate titrant, starch indicator solution, and a teacher's guide and student copy masters.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty