Would you like to log in again?
Determination of the Alkalinity of Water
Supplier: ALDON CORP SE
IS9752
470146-440EA 50 USD InStock
470146-440
Determination of the Alkalinity of Water
Educational Materials Chemistry Educational Materials Mixtures and Solutions Learning Activities
This kit allows analysts to perform a very simple titration to determine the alkalinity and the phenolphthalein of their fresh water sample.
  • Hands-on student activity
  • Students can learn to titrate in the lab or in the field
  • Activity explains water's ability to neutralize acids
  • Lab provides instructions
  • Solutions included
  • Aligned with GLOBE protocols
The titration uses an easily identifiable color transition to indicate the end of the analysis.
The kit contains everything an analyst would need to perform a water analysis either in the laboratory setting or out in the field.
Ordering information: This kit includes plastic pipettes, a reaction vessel, a collection bottle, a measuring tube, titrator, hydrochloric acid titrant, total alkalinity indicator solution, phenolphthalein indicator solution, and an instruction manual.
Delivery information: Ground Delivery Only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty