Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Short Glass Vials with Screw Caps
Supplier: GLASS VIALS INC SE
V2595C-FM-SP V1965C-FM/SP V1760C-FM-SP V1545C-FM/SP V2170C-FM-SP
470146-656CS 574.6 USD InStock
470146-656 470202-764 470146-660 470146-654 470202-766
Short Glass Vials with Screw Caps
Vials
Ideal for a variety of uses.
  • Borosilicate glass with clear screw thread vials
  • Foam-lined polypropylene closed top caps
  • Autoclavable
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty