Would you like to log in again?
AlniMAX II™ Stainless Steel Multipole Magnets
Supplier: SUNDOWN INDUSTRIES SE
CW-010-3PK
470146-710CS 1025.95 USD InStock
470146-710
AlniMAX II™ Stainless Steel Multipole Magnets
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Stronger Than Alnico, But Won’t Attract Another Magnet.
  • Developed as an improved “cow magnet,” these 3" long x 1/2" diameter magnets are ideal for educational use too
  • The special special stainless steel alloy gives them great magnetic strength and remanence, as well as outstanding corrosion resistance
To protect the cow’s stomachs from damage by sharp metal buildup on the ends, the magnets do not have a pole at each end. Instead, many poles are distributed thoroughout, so the magnets attract small metal items all over, but don’t attract or repel another magnet! Check its field out with iron filings or Mag Chips! Package of three.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty