Would you like to log in again?
Heat Solution
Supplier: Worldwide Products
RH36
470146-718CS 416.8 USD InStock
470146-718
Heat Solution
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Unique and Dramatic Change-of-State Demonstrations In a Convenient Pack
  • Eliminate the need for messy chemicals in change-of-state demonstrations with this portable, non-toxic heat pack
  • 13 x 9 cm size
The pack contains a super-cooled and super-saturated solution made of food-grade sodium acetate and water. A stainless 1.7cm steel disk is is also enclosed. Flexing the disk activates two simultaneous chain reactions which induce rapid crystallization. Stored energy is released throughout the solution as it crystallizes. In about five seconds, the pack can reach a temperature of up to 40°C and will stay warm for about two hours. The pack is reusable, simply recharge it in boiling water for about six minutes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty