Would you like to log in again?
Ward's® Syringe-style Micropipettes
Supplier: Ward's
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
11003-010W 11003-003 11002-004 WARD470019-816
470019-814EA 44.3 USD InStock
470019-814 470019-816 470019-918 470146-860
Ward's® Syringe-style Micropipettes
Pipettes Micropipettes
Economical choice for basic applications.
  • Measures 10, 20, 50, 100, and 200 μl
  • Comes with two tips and a tip gauge
  • Optional rack/stand keeps equipment safe between use
Measure and transfer small amounts of liquid with our inexpensive micropipette. Each tip is graduated for accurate measurements, and the built-in stop at 100 μl prevents any overflow into the tip adapter.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty