Would you like to log in again?
Square Bottle, Graduated
Supplier: Dynalon
226295-0500
470147-492CS 349.9 USD InStock
470147-492
Square Bottle, Graduated
Bottles Plastic Bottles
Save valuable shelf space while providing greater materials storage.
  • Graduated plastic square bottles are ideal for long term sample storage
  • Approximate graduations allow reproducible filling without the use of cylinders/beakers
  • Enhanced stability when used with an ergonomic hand grip, allowing greater ease in pouring
  • Screw capped
Convenient square bottle design saves space. The integrity of the seal is insured by an additional internal plug. Hole in square bottle closure for attachment of tag.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty