Would you like to log in again?
Supertek Plastic Hall’s Carriage
Supplier: VWR International
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Supertek
P10507
470148-700CS 648 USD InStock
470148-700
Supertek Plastic Hall’s Carriage
Educational Materials Physics Educational Materials Mechanics Learning Activities
Watch work and energy intercept.
This lightweight, nearly frictionless Hall’s Carriage is perfect for studying the relationship of work and energy. The wide plastic body can carry bulk loads. Oil-free sleeve bearings never need adjustment. Use with our Picket Fence for determining the coefficient of rolling friction. Instructions included. Size: 13 cm L.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty