Would you like to log in again?
Constant Motion Machine
Supplier: VWR International
P10509
470148-714CS 757.45 USD InStock
470148-714
Constant Motion Machine
Educational Materials Physics Educational Materials Mechanics Learning Activities
Students construct simple motion experiments with this machine.
  • Demonstrate the relationship between velocity, displacement and time
  • Compact but powerful
  • 18×10×6.5 cm
  • Runs on two AA batteries
Demonstrate the principles of uniform linear motion using this powerful 'toy'. Moving with a constant speed, this vehicle demonstrates the relationship between velocity, displacement and time. The vehicle can also tow dynamics carts to demonstrate vectors. AA batteries make this a convenient alternative to other vehicles.
Ordering information: Two AA batteries required, but not included.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty