Would you like to log in again?
Inclined Plane Set
Supplier: VWR International
P10805A
470148-758CS 502.25 USD InStock
470148-758
Inclined Plane Set
Educational Materials Physics Educational Materials Mechanics Learning Activities
Present a Wide Variety of Standard Force and Motion Activities.
  • The versatile set makes it easy to investigate acceleration, friction, gravity, and Galileo’s free-fall experiments
  • The inclined plane folds easily for storage with a special hinge, clamps to 45°, and includes a protractor and pulley with a mere 0.04 coefficient of friction
Unlike silk-screened versions, the rugged PVC scale can’t wear off. The all-aluminum construction means it’s virtually student-proof. It comes with Hall’s carriage, pulley, weight pan, solid aluminum plane, and accessories. Illustrated instructions include projects and sample problems.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty