Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Thermometer Rack
Supplier: Shiv Dial Sud & Sons
CT1958
470148-764CS 228.95 USD InStock
470148-764
Thermometer Rack
Thermometer Holders
Convenient and autoclavable.
  • Holds up to 25 thermometers
  • Holds thermometers in a safe vertical position to avoid damage while drying
  • Made of HDPE
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty