Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Demonstration Diffraction Grating

Supplier: VWR International

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

PH30677/A
470148-784CS 7334.65 USD InStock
470148-784
Demonstration Diffraction Grating
Educational Materials Physics Educational Materials Optics Learning Activities
This quality card slide lets you demonstrate and compare the properties of light waves.

  • Viewing Area Length: 9 mm
  • Viewing Area Width: 16 mm
  • Gratings Mounted between Glass Plates
  • Three Diffraction Gratings
  • For Demonstrating Properties of Light Waves

It features three different diffraction gratings: 100, 300 and 600 lines per mm.

The gratings are mounted between glass plates.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty