Would you like to log in again?
Standard Bunsen Burners
Supplier: VWR International
C-1476/3 C-1484 C-1477
470148-792CS 273.4 USD InStock
470148-792 470148-926 470148-654
Standard Bunsen Burners
Heaters Gas Burners
These standard Bunsen burners are practical and economical.
  • Gas Inlet: 5/16" Diameter
  • Mixing Tube: ½" Diameter without Stabilizer
  • Mixing Tube: 7/10" Diameter with Stabilizer
  • Height: 6⅛" without Stabilizer
  • Height: 6⅜" with Stabilizer
They feature an aluminum mixing tube and a sturdy die-cast, nickel-plated base.
With a stabilizer, they have a height of 6⅛".
Neoprene Bunsen Burner Tubing is recommended.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty