Would you like to log in again?
Clamp Pulley, Adjustable
Supplier: VWR International
P10337A
470148-812CS 1264.6 USD InStock
470148-812
Clamp Pulley, Adjustable
Educational Materials Physics Educational Materials Mechanics Learning Activities
This versatile pulley can be used on table edges or at different heights.
  • Nylon Construction
  • Steel Cone Bearing
  • Rod Length: 15 cm
  • Pulley Works on Table Edges or at Different Heights
  • For Experiments Requiring Pulleys to Have Support at Various Heights
It's ideal for experiments that require a pulley to be supported at various heights.
The 40 mm nylon pulley is mounted on steel cone bearings in a metal fork. The fork is attached to a 15 cm rod.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty