Would you like to log in again?
Visual Scientifics Base
Supplier: EISCO SCIENTIFIC LLC
PTBASE
470148-822CS 3876 USD InStock
470148-822
Visual Scientifics Base
Educational Materials Physics Educational Materials Mechanics Learning Activities
This base platform supports the full range of Visual Scientifics activities and accessories.
  • High-contrast grid
  • Magnetic
  • Platform neutral - works with your preferred probeware/video analysis tools
Your students can set up their experiments quickly and easily with this large, sturdy, magnetic base that features threaded junction points for precise probeware mounting. Used in conjunction with Visual Scientifics accessory equipment, this laboratory investigation system offers students an engaging platform and accurate results. Its high-contrast color grid also allows for easy data collection with video analysis systems.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty