Would you like to log in again?
Essential Physics Demo: Faraday's Disk
Supplier: VWR International
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
P41226
470148-896CS 485.85 USD InStock
470148-896
Essential Physics Demo: Faraday's Disk
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Grab and hold students' attention with these interactive and affordable demonstrations that show core physics concepts in action.
  • Demos are available for all fundamental physical science topics
Ideal for jump starting your unit with exciting visual impact, or for reinforcing key concepts after a lesson, these apparatus are quick and easy to set up to save you time in the classroom. Demos are available for all fundamental physical science topics, and include detailed instructions.
Students are used to seeing voltage induced by a coil of wire spun in a magnetic field; now let them see how a solid disk can produce the same results. Potential difference is induced as a 15 cm copper disk is spun through a pair of strong magnets. Required but not included: a multimeter that can measure in the mV range.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty