Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Supertek Acceleration Timers

Supplier: VWR International
Supertek
P10563A P10555A
470148-906CS 4415 USD InStock
470148-906 470148-902
Supertek Acceleration Timers
Timers Mechanical Timers
Calculate acceleration, drag, and friction by measuring the interval between dots.

  • Mechanical timer device
  • 60 Hz (mains power frequency)
  • Timer size: 7L×2W×1¹/₄" H
  • Tape length: 300 m per roll
  • Tape width: 9 mm
  • Carbon disk Ø: 38 mm

This timer works like a clock and produces precisely sixty dots per second on a moving strip of paper. The unit is heavy enough to rest on the edge of a table without clamps and is also designed to clamp to an upright support rod or crossbar. The timer is powered directly by 110 V AC via a 5' 10" cord with an on/off switch.

Ordering information: Timer is supplied with 12 carbon disks and instructions. Tape is required and sold seperately. Not available in Canada.

Order Now
 

ORDER

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty