Would you like to log in again?
High Form Porcelain Crucibles
Supplier: SILICO & CHEMICO PORCELAIN SE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
J240 J238 J237 J232 J239 J241
470149-562CS 139.65 USD InStock
470149-562 470149-078 470149-564 470148-980 470148-954 470148-910
High Form Porcelain Crucibles
Crucibles
These crucibles are similar to wide-form porcelain crucibles but taller.
  • Without covers
  • Max. use temp. is 1050 Celsius
  • Rate of cooling and heating should not increase 200 degrees per hour
They don't have covers.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty