Would you like to log in again?
Liquid Convection Apparatus
Supplier: VWR International
P20318
470148-912CS 2584.7 USD InStock
470148-912
Liquid Convection Apparatus
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Effective Method of Showing Normally Invisible Currents
A rectangular glass tube has an opening at the top in which to pour water and introduce dye, which is available separately. When a corner of the tube is heated, the current is visible in the movement of the dye.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty