Would you like to log in again?
Wheel & Axle
Supplier: VWR International
P10405
470148-942CS 402 USD InStock
470148-942
Wheel & Axle
Educational Materials Physics Educational Materials Mechanics Learning Activities
Show the mechanical advantage of the wheel and axle.
  • The rod can be clamped to a support stand using a right angle clamp for multiple uses!
The pulley is stepped with 35, 50, 65, and 105 mm diameters, each grooved to allow several turns of cord. It is fitted with ball bearings that turn on a steel rod.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty