Would you like to log in again?
Hand Held Color Wheel
Supplier: VWR International
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
P30703A
470149-010CS 348.5 USD InStock
470149-010
Hand Held Color Wheel
Educational Materials Physics Educational Materials Optics Learning Activities
Reveal the Nature of Color and Light
  • When you spin this classic color wheel, all the colors of the rainbow are blended together
  • Challenge students to decipher why colors that are separate when the wheel is still appear to mix when the wheel is in motion
This simple demonstration reveals how light interacts with matter by transmission, absorption, or scattering; and it explains that to see an object, light emitted by or scattered from that object must enter the eye. Size: 9" dia.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty