Would you like to log in again?
Ward’s® Biochemistry Molecular Model Teacher Set, 562 Pieces
Supplier: VWR International
SET00611
470149-022KT 101.1 USD InStock
470149-022
Ward’s® Biochemistry Molecular Model Teacher Set, 562 Pieces
Models Molecular Biology Models
Enhance Instructor-Led Biochemistry Lessons
Instructors can easily demonstrate key biochemistry concepts and chemical structure with this colorful molecular model. Featuring elements most commonly found in biochemistry, this set contains 270 color-coded spheres for carbon, oxygen, hydrogen, nitrogen, sulfur, phosphorus and metal, with holes for inserting rods that represent chemical bonds. Includes 200 short bonds, 30 medium bonds, and 30 long bonds, with link remover tool. Spheres range in diameter from 15mm to 22mm.
Contents:
Qty Element Color Holes Diameter
45 Carbon Black 4 22 mm
25 Carbon Black 3 22 mm
24 Oxygen Red 2 20 mm
10 Oxygen Red 1 15 mm
10 Oxygen Red 2-linear 20 mm
100 Hydrogen White 1 15 mm
10 Hydrogen White 2 15 mm
14 Nitrogen Blue 4 22 mm
12 Nitrogen Blue 3 22 mm
10 Nitrogen Blue 2 20 mm
2 Sulfur Yellow 2 22 mm
6 Phosphorus Purple 4 22 mm
1 Metal Grey 4 22 mm
1 Metal Grey 6 22 mm
30 Medium Bonds Grey
30 Long Bonds Grey
10 Medium Bonds Purple
200 Short Bonds White
Link Remover Tool
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty