Would you like to log in again?
Friction Block Set
Supplier: VWR International
PH10580A
470149-098CS 1715.65 USD InStock
470149-098
Friction Block Set
Educational Materials Physics Educational Materials Mechanics Learning Activities
The Friction Block Set is ideal for simple friction experiments.
  • Ideal for Simple Friction Experiments
  • Instructions Included
  • Attached Mirror
This set consists of a glass mirror with sandpaper affixed to one side and two wooden blocks: one with a hook and the other with an eye. The latter block also has a mirror cemented to one face.
It comes with instructions for use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty