Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Precision Linear Expansion Apparatus

Supplier: VWR International

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

P20047/3 P20047/7 P20047 P20047/5 P20047/1
470149-176CS 1112.75 USD InStock
470149-176 470149-110 470149-042 470149-192 470148-932
Precision Linear Expansion Apparatus
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
The Precision Linear Expansion Apparatus produces accurate determinations of the coefficient of expansion.

  • For Measuring Thermal Expansion of Metals
  • Choice of Four Kinds of Rods: Aluminum, Steel, Copper, and Brass
  • No Electrical Accessories Available
  • Readings Are Repeatable to 0.005 mm
  • Easy-To-Replace Expansion Rods

This rigidly constructed apparatus requires no electrical accessories, and readings are repeatable to 0.005 mm. Expansion rods can be easily replaced without removing stoppers by sliding one rod into the heating jacket that automatically ejects the other rod.

Four expansion rods are available: aluminum, brass, copper, and steel. Rods measure 60 cm x 6.4 mm (1/4") in diameter. A steam generator is required.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty