Would you like to log in again?
Wheatstone Bridge
Supplier: VWR International
P50407A
470149-160CS 2059.35 USD InStock
470149-160
Wheatstone Bridge
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The Wheatstone Bridge is complete with binding posts.
  • One Length of AWG 30 Nickel Silver Wire
  • Scale Graduated in mm
  • Slide Knife-Edge Contact
  • Binding Posts
The apparatus consists of one length of AWG 30 nickel silver wire mounted on a scale graduated in mm. The scale features a slide knife-edge contact.
For known and unknown resistance, two strips of heavy aluminum are mounted in each of the two arms of the bridge.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty