Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vacuum Plate and Pad

Supplier: VWR International

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

PH36018A/1 PH36018A/3
470149-426CS 1324 USD InStock
470149-426 470149-428
Vacuum Plate and Pad
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
This vacuum plate and pad are the perfect tools for vacuum experiments.

Here’s the perfect plate for your experiments requiring a vacuum situation. It works with bell jars and a vacuum pump – plus, no messy vacuum wax is needed. This handy plate for any lab features a 50 mm center hole, safety screw plug, and stopcock with hose nipple for vacuum tubing of 1/4" ID.

  • No Messy Vacuum Wax Needed
  • Air-Tight Contact
  • Ideal with Bell Jars and Vacuum Pumps
  • Center Hole: 50 mm
  • Safety Screw Plug

They work with bell jars and vacuum pumps, and no messy vacuum wax is needed. The handy plate features a 50 mm center hole, safety screw plug, and stopcock with hose nipple for vacuum tubing of 1/4" ID.

The top plate measures 11" in diameter. It's made of cast iron.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty