Would you like to log in again?
Water Turbine with Dynamo
Supplier: VWR International
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
P20445
470149-432EA 90.6 USD InStock
470149-432
Water Turbine with Dynamo
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Create Light with the Power of Water
  • User Friendly
  • High Quality
Demonstrate how the power of water can be used to produce clean energy. Water pressure from your sink faucet churns the turning blades, creating enough energy to power an LED bulb when the optimum water pressure is reached. The open design of this water turbine allows students to clearly observe the motion of the blades and the energy-producing action of the dynamo. Required but not included are a hose and water source.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty