Would you like to log in again?
Closed Case Form Electroscope
Supplier: VWR International
Avg. Ratings: 4.0 (1 Review)
REVIEWS (1)
Total Ratings: 1
Avg. Ratings: out of 5
Sort all by:
P45258S P45258
470149-480CS 1042.55 USD InStock
470149-480 470148-720
Closed Case Form Electroscope
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Closed Case Form Electroscope is enclosed, preventing air currents from affecting the deflection of the leaves.
  • Booklet of Six Leaves
  • Instructions
  • Gold and Aluminum Leaf Construction
  • Enclosed Electroscope
  • Leaves Pivot when Charged
Leaves are a combination of gold and aluminum developed to provide strength without sacrificing sensitivity. Each leaf is suspended from two small hooks for simple installation, and the leaves pivot, rather than bend, when charged.
The metal case eliminates deflections caused by charges residing on glass walls. An insulating plastic sleeve holds the leaf-support rod in place. Front and rear sides of the box are made of a 10 cm square glass pane to allow visibility and shadow projection.
Ordering information: This kit includes a booklet of six leaves and instructions.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (1)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty