Would you like to log in again?
Economy Rod-Mounted Pulley
Supplier: VWR International
P10317A
470149-504CS 1977.25 USD InStock
470149-504
Economy Rod-Mounted Pulley
Educational Materials Physics Educational Materials Mechanics Learning Activities
Excellent for Introductory Mechanics Studies
  • Low friction cone bearings ensure accurate, repeatable results
Consists of a 10cm-long rod securely fastened to a durable nylon fork. The nylon sheave is 4.5cm in diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty