Would you like to log in again?
Bulb socket board
Supplier: VWR International
PH60122A/1-B PH60122A/3
470149-514CS 880.5 USD InStock
470149-514 470149-512
Bulb socket board
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
This solid socket board holds light bulbs for circuit experiments.
  • Choice of Three or Five-Socket Sizes
  • Removable Shunt Wires Featured on Five-Socket Size
  • Terminals Featured on Three-Socket Board
  • Plastic Bases with Rubber Feet
  • Small Incandescent Bulbs
It's available in three or five-socket sizes. Terminals on three-socket board let users connect lamps in series or parallel, and five-socket board features removable shunt wires for the same purpose.
All units have plastic bases with rubber feet, and small incandescent bulbs are included.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty