Would you like to log in again?
Speed of a Bubble Set
Supplier: VWR International
PH10639A
470149-527CS 1760.5 USD InStock
470149-527
Speed of a Bubble Set
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
The Speed of a Bubble Set connects the mathematical concept of slope to the physical science concept of velocity.
  • Instructions Included
  • Three Tubes Contain Three Fluids for Bubbles
  • Different Factors Affect Bubbles' Speed
  • Students Track Bubbles' Time through Tubes
  • Purpose for Students: to Link the Concept of Slope to That of Velocity
It consists of three rugged, sealed, and transparent plastic tubes containing colored fluids of three different viscosities. The bubble in each tube rises at a constant speed, depending on the type of fluid, the angle at which the tube is inclined, and the temperature.
Students track the time it takes for an air bubble to move from the bottom to the top of the different-colored tubes. Instructions are included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty