Would you like to log in again?
Helical Spring Set
Supplier: STANLEY SPRING & STAMPING
6645601B&L
470149-836PK 46.95 USD InStock
470149-836
Helical Spring Set
Educational Materials Physics Educational Materials Mechanics Learning Activities
This set consists of a carefully selected steel spring, a mass of 0.5 kg with a hook on both ends, and a mass of 1 kg with a single hook.
The spring has hooks on both ends and is approximately 20 cm long.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty