Would you like to log in again?
Small Animal Housing
Supplier: MARYLAND PLASTICS INC SE
E-0494B-B
470149-838CS 518.35 USD InStock
470149-838
Small Animal Housing
Animal Supplies Animal Cages
Chemical and solvent-resistant polypropylene cages for housing and breeding mice, rats or hamsters.
  • Autoclavable to 230 °C
  • Feature runners on the bottom for air circulation
  • Stainless covers with a hinged divider separating the food hopper and space for the water bottle
Covers are available separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty