Would you like to log in again?
Immunotechnology Activity Kit
Supplier: GENO TECHNOLOGY SE
BE-506
470150-030KT 200.09 USD InStock
470150-030
Immunotechnology Activity Kit
Educational Materials Chemistry Educational Materials Chromatography Learning Activities
Hands-on Study of Immunoprecipitation
This kit contains all the reagents necessary to immunoprecipitate a specific protein from a complex sample. This hands-on activity focusing on immunoprecipitation permits the purification of specific proteins for which an antibody has been raised. The kit includes enough materials for six groups of four students to complete the experiment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty