Would you like to log in again?
GPI Anatomicals® Introductory Artery Model
Supplier: GPI ANATOMICALS
2600
470150-760EA 118.55 USD InStock
470150-760
GPI Anatomicals® Introductory Artery Model
Models Human Biology Models
Arterial Structure Represented With Cholesterol Deposits
Ideal for medical studies as well as anatomy classes, the 100X artery model is sectioned to show the arterial wall and cellular structure, as well as the partial occlusion of one branch by cholesterol. The model is mounted on a base and can be removed for closer examination.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty