Would you like to log in again?
Fingertip Safety Tape
Supplier: GENERAL BANDAGES INC.
41408-3/4
470150-976CS 85.49 USD InStock
470150-976
Fingertip Safety Tape
First Aid Supplies First Aid Kit Components
Protect your fingers without gloves.
This self-adhering tape maintains an unimpaired sense of touch, yet prevents hand and finger injuries from sharp edges, hot surfaces, or rough materials.
Made from pure surgical grade cotton gauze coated with natural latex rubber. Roll measures 1.9 cm wide × 27 m long.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty