Would you like to log in again?
Ward's® Etched Petrographic Microscope Slides
2795-602765
470151-066CS 462.8 USD InStock
470151-066
Ward's® Etched Petrographic Microscope Slides
Slides Microscope Slides
Save Time and Prep Work
These slides are pre-etched to increase rock adhesion to the slide. Measures 1" x 2"; 27 x 46 mm. Edges are beveled, ground, and polished. Thickness: 1.2 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty