Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
MyChron Stopwatch
Supplier: DANTONA INDUSTRIES INC SE
460262 460261 WARD470151-280 UL389
470175-282BX 2377 USD InStock
470175-282 470175-286 470031-564 470151-280
MyChron Stopwatch
Timers Digital Stopwatches
The MyChron stopwatch times lab activities with precision.
  • Large buttons
  • Color-coded
  • A comparable replacement is the Energizer no 389 (SR1130W), 1.5 V or similar type battery
The MyChron Stopwatch has large, color-coded buttons.
It features a lap mode and is accurate to the hundredth of a second.
Packaging: Comes with one G10-A button battery.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty