Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Nickel Metal Hydride Rechargeable Batteries
Supplier: RAYOVAC CORPORATION
PL713-2 GENB PL715-4 GENB
470152-292CS 179.1 USD InStock
470152-292 470152-296
Nickel Metal Hydride Rechargeable Batteries
Batteries
Nickel Metal Hydride or NiMH batteries are considered better than NiCad batteries as they are less likely to exhibit the 'memory' effect, have a higher storage capacity (25%), and there is no toxic cadmium to dispose of when they finally wear out.
NiMH batteries last an average 6000 hours and can keep 80% of their charge at −20°. You can also recharge these AA and AAA batteries with the charger, available separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty