Would you like to log in again?
Brine Shrimp Eggs
Supplier: VWR
WARD470152-880
470152-880PK 5.95 USD InStock
470152-880
Brine Shrimp Eggs
Specimens Live Specimens Aquaria - Living Material
Place these eggs in a brine solution for two days in order to make the shrimp (Artemia) hatch. Our special processing technique allows you to store the eggs for nearly indefinite periods of time. 7 grams.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty